Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567313 Supplier Invitrogen™ Supplier No. PA567313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLEVHFVGDDD VLNHILDREG GAKLKKERAQ LLVNPKKIIK KPEYDLEEDD QEVLKDQNYV EIMGRDVQES LKNGSATGGG NKVYSFQNRK HSEKMAK Highest antigen sequence identity to the following orthologs - mouse 69%, rat 70%.

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC2
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ORC2
Gene Accession No. Q13416
Gene Alias AU041563; Orc2; ORC2L; origin recognition complex protein 2 homolog; origin recognition complex subunit 2; origin recognition complex, subunit 2; origin recognition complex, subunit 2 homolog; origin recognition complex, subunit 2-like; zgc:153041
Gene Symbols ORC2
Host Species Rabbit
Immunogen Recombinant Human ORC2. Recombinant protein control fragment (Product #RP-105586).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4999
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.