Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598032
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598032 Supplier Invitrogen™ Supplier No. PA598032
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ORC4 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The origin recognition complex (ORC) is a highly conserved six subunit protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. This gene encodes a subunit of the ORC complex. Several alternatively spliced transcript variants, some of which encode the same protein, have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ORC4
Gene Accession No. O43929
Gene Alias HSORC4; mMmORC4; Orc4; ORC4L; Orc4P; Origin recognition complex subunit 4; origin recognition complex, subunit 4; origin recognition complex, subunit 4 homolog; origin recognition complex, subunit 4-like
Gene Symbols ORC4
Host Species Rabbit
Immunogen Recombinant Human Origin recognition complex subunit 4 protein (187-436AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5000
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.