Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121127 Supplier Invitrogen™ Supplier No. PA5121127
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, MCF7, Mouse brain. The target is usually found in the following locations: Nucleus. Immunogen sequence: IAVIGLTCRL DILELLEKRV KSRFSHRQIH LMNSFGFPQY VKIFKEQLSL PAEFPDKVFA EKWNENVQYL SEDRSVQEVL QKHFNISKNL RSLHMLLMLA LNRVTASHPF MTAVDLMEAS QLCSMDSKAN IVHGLSVLEI CLIIAMKHLN DIYEEEPFNF QMVYNEFQKF VQRKAHSVYN FEKPVVMKAF EHLQQLELIK PMERTSGNSQ REYQLMKLLL DNTQIMNALQ KYPNCPTDVR QWATSSLSWL.

The origin recognition complex (ORC) is a highly conserved six subunit protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. This gene encodes a subunit of the ORC complex. Several alternatively spliced transcript variants, some of which encode the same protein, have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ORC4
Gene Accession No. O43929, O88708
Gene Alias HSORC4; mMmORC4; Orc4; ORC4L; Orc4P; Origin recognition complex subunit 4; origin recognition complex, subunit 4; origin recognition complex, subunit 4 homolog; origin recognition complex, subunit 4-like
Gene Symbols ORC4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 187-436 of human ORC4L (NP_002543.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26428, 5000
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.