Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORC5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100887 Supplier Invitrogen™ Supplier No. PA5100887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ORC5.

Component of the origin recognition complex (ORC) that binds origins of replication. DNA-binding is ATP-dependent. The specific DNA sequences that define origins of replication have not been identified yet. ORC is required to assemble the pre- replication complex necessary to initiate DNA replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC5L
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ORC5
Gene Accession No. O43913, Q9WUV0
Gene Alias AL033327; MmORC5; ORC5; Orc5l; ORC5P; ORC5T; origin recognition complex 5; origin recognition complex subunit 5; origin recognition complex, subunit 5; origin recognition complex, subunit 5 homolog; origin recognition complex, subunit 5-like; PPP1R117; protein phosphatase 1, regulatory subunit 117
Gene Symbols ORC5
Host Species Rabbit
Immunogen A synthesized peptide derived from human ORC5(Accession O43913), corresponding to amino acid residues H227-Q277.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26429, 5001
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.