Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120914 Supplier Invitrogen™ Supplier No. PA5120914
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Jurkat, SW620, PC-3, HeLa, Mouse brain, Rat brain. The target is usually found in the following locations: Nucleus. Immunogen sequence: MGSELIGRLA PRLGLAEPDM LRKAEEYLRL SRVKCVGLSA RTTETSSAVM CLDLAASWMK CPLDRAYLIK LSGLNKETYQ SCLKSFECLL GLNSNIGIRD LAVQFSCIEA VNMASKILKS YESSLPQTQQ VDLDLSRPLF TSAALLSACK ILKLKVDKNK MVATSGVKKA IFDRLCKQLE KIGQQVDREP GDVATPPRKR KKIVVEAPAK EMEKVEEMPH KPQKDEDLTQ DYEEWKRKIL ENAASAQKAT AE.

The origin recognition complex (ORC) is a highly conserved six subunit protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. Gene silencing studies with small interfering RNA demonstrated that this protein plays an essential role in coordinating chromosome replication and segregation with cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Orc6
Gene Accession No. Q9WUJ8, Q9Y5N6
Gene Alias 6720420I10Rik; ORC 6; ORC6; ORC6L; origin recognition complex subunit 6; origin recognition complex, subunit 6; origin recognition complex, subunit 6-like
Gene Symbols Orc6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-252 of human ORC6 (NP_055136.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23594, 291927, 56452
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.