Learn More
ORC6 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301492AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
The origin recognition complex (ORC) is a highly conserved six subunit protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. It has been shown that this protein and and ORC1L are loosely associated with the core complex consisting of ORC2L, -3L, -4L and -5L. Gene silencing studies with small interfering RNA demonstrated that this protein plays an essential role in coordinating chromosome replication and segregation with cytokinesis.Specifications
| ORC6 | |
| Polyclonal | |
| Unconjugated | |
| ORC6 | |
| ORC6L; origin recognition complex, subunit 6 homolog-like (yeast); ORC6; origin recognition complex subunit 6; origin recognition complex, subunit 6-like (yeast); origin recognition complex, subunit 6 (yeast homolog)-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23594 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9Y5N6 | |
| ORC6 | |
| Between 200 and 230 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.