Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORC6L Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074619AF700
Description
ORC6L Polyclonal antibody specifically detects ORC6L in Human samples. It is validated for Western BlotSpecifications
ORC6L | |
Polyclonal | |
Western Blot | |
ORC6Lorigin recognition complex, subunit 6 homolog-like (yeast), origin recognition complex subunit 6, origin recognition complex, subunit 6, origin recognition complex, subunit 6 like (yeast) | |
The immunogen recognized by this antibody maps to a region between residue 200 and 230 of human origin recognition complex subunit 6 using the numbering given in entry NP_055136.1 (GeneID 23594). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
23594 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction