Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Orexin A, Human, Bovine, Rat, Mouse

Catalog No. aaj65378exd
Click to view available options
Quantity:
0.5 mg

3561.09 g/mol

appetite regulation, wakefulness, locomotor activity, hypothalamic-pituitary-adrenal activity, and pain thresholds. Orexin signaling is activated by nutrient depletion, causing an increase in food intake by delaying the signals of satiety. It activates the orexin-1 and -2 receptors with equal affinity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Species Bovine, Mouse, Rat
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 3561.09
Product Type Orexin A, human, bovine, rat, mouse
Quantity 0.5 mg
Regulatory Status RUO - research use only

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.