Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orexin A/Hypocretin-1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199426 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017695 0.1 mL
NB10065204 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017695 Supplier Novus Biologicals Supplier No. NB100652040.1ML
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Orexin A/Hypocretin-1 Polyclonal specifically detects Orexin A/Hypocretin-1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Orexin A/Hypocretin-1
Applications Immunohistochemistry (Frozen), ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:200-1:800, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunohistochemistry-Frozen 1:10-1:500
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Accession No. O43612
Gene Alias hypocretin (orexin) neuropeptide precursor, NRCLP1, orexin, OXHcrt, PPORX, PPOXHypocretin
Gene Symbols HCRT
Host Species Sheep
Immunogen Synthetic peptide corresponding to amino acids 2-14 of human orexin A peptide, conjugated to KLH.
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3060
Test Specificity NB100-65204 is specific for human orexin A.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.