Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orexin A/Hypocretin-1 Antibody (MM0500-8G22) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50

Specifications

Antigen Orexin A/Hypocretin-1
Clone MM0500-8G22
Concentration LYOPH
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018914
SDP
View Documents
Novus Biologicals
NBP2118330.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP211833
SDP
View Documents
Novus Biologicals
NBP211833
0.1 mg Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Orexin A/Hypocretin-1 Monoclonal specifically detects Orexin A/Hypocretin-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Orexin A/Hypocretin-1
LYOPH
Unconjugated
Mouse
Cancer, GPCR, Neuronal Cell Markers, Neuroscience
O43612
3060
IgG1
Protein G purified
MM0500-8G22
Monoclonal
Purified
RUO
Human
hypocretin (orexin) neuropeptide precursor, NRCLP1, orexin, OXHcrt, PPORX, PPOXHypocretin
HCRT
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.