Learn More
Invitrogen™ Orexin A Polyclonal Antibody

Description
Predicted to react with bovine, chimpanzee, dog, horse, mouse, pig, rabbit, rat and rhesus monkey based on sequence homology. Hypothalamus tissue can be used as a positive control. Sheep anti Human orexin A antibody recognizes human orexin A, an excitatory neuropeptide linked to alertness and wakefulness in humans.
Specifications
Specifications
| Antigen | Orexin A |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | HCRT |
| Gene Accession No. | O43612 |
| Gene Alias | HCRT; Hcrt1; Hcrt2; Hypocretin; Hypocretin (orexin) neuropeptide; hypocretin neuropeptide precursor; hypocretin-1; Hypocretin-2; NRCLP1; orexin; orexin A; orexin B; orexin-A; Orexin-B; ORX; OX; PPORX; PPOX; prepro-orexin |
| Gene Symbols | HCRT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.