Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Orexin A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570436
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 93%; Mouse: 100%; Pig: 93%; Rabbit: 93%; Rat: 100%; Sheep: 93%.
This gene encodes a hypothalamic neuropeptide precursor protein that gives rise to two mature neuropeptides, orexin A and orexin B, by proteolytic processing. Orexin A and orexin B, which bind to orphan G-protein coupled receptors HCRTR1 and HCRTR2, function in the regulation of sleep and arousal. This neuropeptide arrangement may also play a role in feeding behavior, metabolism, and homeostasis.
Specifications
Orexin A | |
Polyclonal | |
Unconjugated | |
HCRT | |
HCRT; Hcrt1; Hcrt2; Hypocretin; Hypocretin (orexin) neuropeptide; hypocretin neuropeptide precursor; hypocretin-1; Hypocretin-2; NRCLP1; orexin; orexin A; orexin B; orexin-A; Orexin-B; ORX; OX; PPORX; PPOX; prepro-orexin | |
Rabbit | |
Affinity chromatography | |
RUO | |
25723 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O55232 | |
HCRT | |
synthetic peptide directed towards the middle region of rat Orexin A. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction