Learn More
Invitrogen™ Orexin B Polyclonal Antibody

Description
PA1-9584 detects Orexin B in human, mouse, and rat samples. PA1-9584 has been successfully used in ELISA procedures. PA1-9584 immunogen is a synthetic peptide corresponding to amino acids 1-14 of human Orexin B.
Specifications
Specifications
| Antigen | Orexin B |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Hcrt |
| Gene Accession No. | O43612, O55232, O55241 |
| Gene Alias | HCRT; Hcrt1; Hcrt2; Hypocretin; hypocretin (orexin) neuropeptide; hypocretin neuropeptide precursor; Hypocretin-1; Hypocretin-2; NRCLP1; orexin; orexin A; orexin B; orexin-A; Orexin-B; ORX; Ox; PPORX; Ppox; prepro-orexin |
| Gene Symbols | Hcrt |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.