Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Orexin R2/HCRTR2 Antibody (456738), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5246G
Description
Orexin R2/HCRTR2 Monoclonal antibody specifically detects Orexin R2/HCRTR2 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Orexin R2/HCRTR2 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
456738 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
hypocretin (orexin) receptor 2, Hypocretin receptor type 2, orexin receptor type 2, Ox-2-R, Ox2-R, OX2R | |
NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2, Met1-Trp444, Accession # AAC39602 | |
0.1 mL | |
Adaptive Immunity, Cancer, GPCR, Immunology, Neuronal Cell Markers, Neuroscience | |
3062 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction