Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Orexin receptor 1+2 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1095R

Description
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene encodes a G-protein coupled receptor that binds orexin A and orexin B.Specifications
Orexin receptor 1+2 | |
Polyclonal | |
Unconjugated | |
HCRTR1 | |
HCRTR1; HCRTR2; Hctr1; hypocretin (orexin) receptor 1; hypocretin (orexin) receptor 2; hypocretin receptor 1; hypocretin receptor 2; Hypocretin receptor type 1; Hypocretin receptor type 2; hypocretin receptor-1; mOX2aR; mOX2bR; Mox2r; mOXR2; orexin receptor 1; orexin receptor 2; Orexin receptor type 1; Orexin receptor type 2; orexin receptor-1; orexin receptor-2; orexin type-2 receptor; Ox1r; Ox-1-R; ox1-R; OX2R; Ox-2-R; ox2-R | |
Rabbit | |
Protein A | |
RUO | |
230777, 25593, 25605, 3061, 3062, 387285 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O43613, O43614, P56718, P56719, P58307, P58308 | |
HCRTR1, Hcrtr2 | |
KLH conjugated synthetic peptide derived from human Orexin receptor 1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction