Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Orexin Receptor 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142162
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142162 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142162 Supplier Invitrogen™ Supplier No. PA5142162
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene (HCRTR2) encodes a G-protein coupled receptor that binds orexin A and orexin B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Orexin Receptor 1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene HCRTR1
Gene Accession No. O43613
Gene Alias Hcrt receptor 1; Hcrtr1; Hctr1; hypocretin (orexin) receptor 1; hypocretin receptor 1; hypocretin receptor type 1; hypocretin receptor-1; Orexin a receptor; orexin receptor 1; orexin receptor type 1; orexin receptor-1; Orexin-1 receptor; OX1 receptor; OX1R; ox-1-R; ox1-R; OX-R1
Gene Symbols HCRTR1
Host Species Goat
Immunogen Peptide with sequence C-RNWKRPSDQLGD (aa 256-267).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3061
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.