Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Orexin Receptor 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533838
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533838 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533838 Supplier Invitrogen™ Supplier No. PA533838
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%) Dog, Rabbit, Pig (93%).

The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene (HCRTR2) encodes a G-protein coupled receptor that binds orexin A and orexin B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Orexin Receptor 1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene HCRTR1
Gene Accession No. O43613, P56718
Gene Alias Hcrt receptor 1; Hcrtr1; Hctr1; hypocretin (orexin) receptor 1; hypocretin receptor 1; hypocretin receptor type 1; hypocretin receptor-1; Orexin a receptor; orexin receptor 1; orexin receptor type 1; orexin receptor-1; Orexin-1 receptor; OX1 receptor; OX1R; ox-1-R; ox1-R; OX-R1
Gene Symbols HCRTR1
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from 3rd cytoplasmic domain of human HCRTR1 / Orexin Receptor 1.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25593, 3061
Target Species Human, Rat, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.