Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Orexin Receptor 2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142719
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein binds the hypothalamic neuropeptides orexin A and orexin B. A related gene encodes a G-protein coupled receptor that selectively binds orexin A.
Specifications
| Orexin Receptor 2 | |
| Polyclonal | |
| Unconjugated | |
| HCRTR2 | |
| HCRTR1; Hcrtr2; hypocretin (orexin) receptor 2; hypocretin receptor 2; hypocretin receptor type 2; mOX2aR; mOX2bR; Mox2r; mOXR2; orexin receptor 1; orexin receptor 2; orexin receptor type 2; orexin receptor-1; orexin receptor-2; orexin type-2 receptor; OTTHUMP00000039969; OX1R; ox2r; ox-2-R; ox2-R | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 387285 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P58308 | |
| HCRTR2 | |
| Peptide with sequence C-KSLTTQISNFDNISK (aa 405-419). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction