Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Orexin Receptor 2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518818
Description
This antibody is predicted to react with bovine, canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein binds the hypothalamic neuropeptides orexin A and orexin B. A related gene encodes a G-protein coupled receptor that selectively binds orexin A.
Specifications
Orexin Receptor 2 | |
Polyclonal | |
Unconjugated | |
HCRTR2 | |
HCRTR1; Hcrtr2; hypocretin (orexin) receptor 2; hypocretin receptor 2; hypocretin receptor type 2; mOX2aR; mOX2bR; Mox2r; mOXR2; orexin receptor 1; orexin receptor 2; orexin receptor type 2; orexin receptor-1; orexin receptor-2; orexin type-2 receptor; OTTHUMP00000039969; OX1R; ox2r; ox-2-R; ox2-R | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
3062 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43614 | |
HCRTR2 | |
Synthetic peptide sequence (SGTKLEDSPPCRN) corresponding to the N-terminus amino acids of HCRTR2 (aa 2-15). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction