Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORF0 Rabbit anti-Primate, Polyclonal, MilliporeSigma™

Catalog No. ABE289525UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2895-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE2895-25UL Supplier MilliporeSigma Supplier No. ABE289525UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-ORF0 Antibody is a highly specific rabbit polyclonal antibody, that targets and has been tested in Immunoprecipitation, Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORF0
Applications Immunoprecipitation, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ORF0
Host Species Rabbit
Immunogen KLH-conjugated linear peptides corresponding to ORF0 amino acid residues 20-34, 33-49, and 50-65.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody recognizes primate-specific open reading frame 0 (ORF0).
Target Species Primate
Content And Storage Stable for 1 year at -20°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.