Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orf2 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992084
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2084 25 μL
50-199-2083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2084 Supplier Rockland Immunochemicals Supplier No. 600401GT3S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Orf2 is directed against mouse ORF2 near a c-terminus position. A BLAST analysis was used to suggest reactivity with this protein from human and feline based on 100% homology for the immunogen sequence.

The multi-functional nuclear protein ORF2p is involved in reverse transcriptase and endonuclease processes. ORF2 elements are epigenetically silenced throughout embyronic development and it is suggested that mutations of L1 would result in detrimental de novo insertions into the genome, negatively affecting gene expression. L1 elements are critical components in the mammalian genome, and much is still not known regarding these proteins. Studies implicate that line-1 is activated in fetal oocytes and L1ORF2p may contribute to the eradication or selection of the oocytes.

Specifications

Antigen Orf2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.