Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORF73/HHV8 Antibody (HHV8/3633R), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308397H
Description
ORF73/HHV8 Monoclonal antibody specifically detects ORF73/HHV8 in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
ORF73/HHV8 | |
Monoclonal | |
HRP | |
HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV | |
Recombinant protein corresponding to the latent nuclear antigen 1 molecule of ORF73/HHV8 | |
100 μg | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
935114 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HHV8/3633R | |
PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction