Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORF73/HHV8 Antibody (LN53), mFluor Violet 450 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122767MFV450
Description
ORF73/HHV8 Monoclonal antibody specifically detects ORF73/HHV8 in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), ImmunoassaySpecifications
ORF73/HHV8 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Virus | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
LN53 | |
Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoassay | |
HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV | |
Recombinant protein corresponding to the latent nuclear antigen-1 molecule of HHV8. | |
0.1 mL | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
935114 | |
Store at 4C in the dark. | |
IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction