Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORF73/HHV8 Mouse anti-Nonspecies specific, Clone: HHV8/3606, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen ORF73/HHV8
Clone HHV8/3606
Concentration 0.2 mg/ml
Dilution ELISA 1-5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123092
SDP
View Documents
Novus Biologicals
NBP30780920UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

ORF73/HHV8 Monoclonal specifically detects ORF73/HHV8 in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

ORF73/HHV8
0.2 mg/ml
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
10 mM PBS with 0.05% BSA
935114
IgG1 κ
Protein A or G purified
HHV8/3606
ELISA 1-5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Non-species specific
HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV
Recombinant protein corresponding to the latent nuclear antigen 1 molecule of ORF73/HHV8
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.