Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Org 24598, >98%
GREENER_CHOICESDP

Catalog No. 444750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-475-0 50 mg
44-471-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-475-0 Supplier R&D Systems™ Supplier No. 4447/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of GlyT1; derivative of Org 24598 (Cat. No. 4035).

Potent and selective inhibitor of the glial glycine transporter GlyT1b (IC50 = 6.9 nM). Displays negligible activity at GlyT-2, adrenoreceptors, dopamine, 5HT receptors and noradrenaline, dopamine, 5HT and GABA transporters (pIC50 < 4). Active in vivo.

Chemical Identifiers

CAS 372198-97-5
Molecular Formula C19H20F3NO3

Specifications

Quantity 50 mg
Solubility Soluble to 10mM in water with gentle warming and to 100mM in DMSO
Product Line Tocris
Formula Weight 367.36
Purity >98%
Molecular Weight (g/mol) 367.368
Chemical Name or Material N-Methyl-N-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]-glycine
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.