Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Org 25543 hydrochloride
GREENER_CHOICESDP

Catalog No. 478250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-825-0 50 mg
47-8210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-825-0 Supplier R&D Systems™ Supplier No. 4782/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective GlyT2 inhibitor

Potent and selective glycine transporter type 2 (GlyT2) inhibitor (IC50 = 16 nM for hGlyT2). Displays no activity at GlyT1 or 56 other common biological targets (≥ 100 μM), in a glycine uptake assay in CHO cells. Ameliorates mechanical allodynia after partial sciatic nerve ligation injury in mice.

Chemical Identifiers

CAS 495076-64-7
Molecular Formula C24H33ClN2O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 448.98
Purity >99%
Molecular Weight (g/mol) 448.988
Chemical Name or Material Org 25543 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.