Learn More
Organic Carbon Standard, 1500 ppm C, Reagents
Specifications
Specifications
| Chemical Name or Material | Organic Carbon Standard, 1500 ppm C |
| Packaging | Poly Bottle |
| CAS | 7732-18-5 |
| Specific Gravity | 1.0049 |
| Physical Form | Liquid |
| Color | Colorless |
| pH | <2 |
| Boiling Point | 100°C |
| For Use With (Equipment) | Water Analyzer |
| Solubility | Completely miscible |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.