Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Organic Carbon Standard, 1500 ppm C, Reagents

Catalog No. p-200021648
Click to view available options
Quantity:
500 mL
1 L
4 L

Organic Carbon Standard, 1500 ppm C, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Organic Carbon Standard, 1500 ppm C
Packaging Poly Bottle
CAS 7732-18-5
Specific Gravity 1.0049
Physical Form Liquid
Color Colorless
pH <2
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Water Analysis
Quantity 1 L
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.