Learn More
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Odor | Odorless |
| Assay Percent Range | 5, 000 ± 10ppm |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Solubility | Infinitely soluble in water |
| Type | Standard |
| Solvent or Matrix | Water |
| Quantity | 4 L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
