Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Organochlorine Pesticide Mix AB No. 3, Restek
SDP

Catalog No. 06719406 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-406 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-406 Supplier Restek Corporation Supplier No. 32415
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Organochlorine Pesticide Mix AB No. 3
Color Colorless
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL each in hexane:toluene (1:1)
Aldrin
α-BHC
β-BHC
δ-BHC
γ-BHC (lindane)
cis-Chlordane
trans-Chlordane
4,4'-DDD
4,4'-DDE
4,4'-DDT
Dieldrin
Endosulfan I
Endosulfan II
Endosulfan sulfate
Endrin
Endrin aldehyde
Endrin ketone
Heptachlor
Heptachlor epoxide (isomer B)
Methoxychlor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.