Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDVOTEK™ Origami Organelles Amino Acids Model

Shop All EDVOTEK Products


Click to view available options
Grade:
4 to 12
Description
- Amino acids are the basic units of proteins that are essential for life.
- Their sequence in proteins is encoded by the genetic code of DNA.
- They have two functional groups - carboxylic acid and an amino group.
- Each amino acid also has a unique side chain (or R group).
- Amino acids are easy to understand with our unique molecular models that show how electrons behave during bond formation.
- Your students first build their easy to make paper models of atoms and electrons.
- They use these to form organic molecules by sharing electrons between the atoms.
- The differently colored electrons make it easy to see how the bonds are formed.
- The models can then be used to show the structure of the 20 naturally occurring amino acids, their physical properties and the formation of peptide bonds.
- Clear instructions are provided in the PowerPoint presentation.
- Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.
- Dimensions: Model atom diameter 10 cm (4 in.)
Specifications
Specifications
Grade | 4 to 12 |
Format | Model |
Languages | English |
Subject | Acids and Bases |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction