Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Cell Structure Model
SDPSCI_ED

Catalog No. S05723 Shop All EDVOTEK Products
Click to view available options
Grade:
4 to 12
This item is not returnable. View return policy
This item is not returnable. View return policy

Show the difference between animal, plant, bacterial, and fungal cells with the Origami Organelles Cell Structure Model.

  • Show what makes animal, plant, bacterial and fungal cells different from each other with our easy-to-make models.
  • To function, they have different combinations of subcellular structures (organelles).
  • Your students make models out of the colorful parts of four different cells.
  • Utilize the model to look at what the subcellular structures do and compare and contrast the different types of cells.
  • The model covers the following: nucleus, chromosome, mitochondrion, chloroplast, vacuole, cell membrane, cell wall, cytoplasm, ribosomes, plasmids, and flagella.
  • Takes just 30 minutes to build the model.
  • Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.
  • Dimensions: Bacterial Cell length: 11 cm (4 in.)
  • Download includes: instructions, model templates, and classroom PowerPoint presentation
  • All you need: paper, scissors, and tape

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology - Cells
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.