Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDVOTEK™ Origami Organelles ELISA Model

Shop All EDVOTEK Products


Click to view available options
Grade:
4 to 12
Description
- Early detection of an infectious disease such as coronavirus is crucial to reducing the spread of such diseases.
- This model shows how the important medical test, enzyme-linked immunosorbent assay (ELISA), is used to detect patients who have a virus or who have previously been exposed to a virus.
- ELISA relies on the use of monoclonal and polyclonal antibodies that are used together to amplify a small signal.
- The model shows how primary antibodies, secondary antibodies, and detection works in direct ELISA, indirect ELISA and sandwich ELISA.
- Time to build model takes about 30 minutes.
- Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.
- Dimensions: 8 cm x 5 cm (3 in. x 2 in.)
- Download includes: instructions, color and mono model templates, and classroom PowerPoint file
- All you need: paper, scissors, tape
Specifications
Specifications
Grade | 4 to 12 |
Format | Model |
Languages | English |
Subject | Biology - Life Science |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction