Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDVOTEK™ Origami Organelles Enzymes Model

Shop All EDVOTEK Products


Click to view available options
Grade:
4 to 12
Description
- This easy to make model gives your students a way to visualize how enzymes catalyze biochemical reactions inside our cells.
- After making a model, use it to look at how enzymes speed up reactions of substrates in their active sites to produce products.
- Then use the model to see how inhibitors work and how enzymes are regulated.
- Covers: enzyme substrate interaction, active site, catalytic cycle, competitive inhibitors, non-competitive inhibitors, allosteric activators, allosteric inhibitors, and cooperativity.
- Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.
- Dimensions: 11 cm x 13 cm (4 in. x 5 in.)
- Download includes: instructions, color and mono model templates, and classroom PowerPoint presentation.
- All you need: paper, scissors, and tape.
Specifications
Specifications
Grade | 4 to 12 |
Format | Model |
Languages | English |
Subject | Biology - Life Science |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction