Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Frog Dissection Model
SDPSCI_ED

Catalog No. S05741 Shop All EDVOTEK Products
Click to view available options
Grade:
4 to 12
This item is not returnable. View return policy
This item is not returnable. View return policy

Dissect a paper frog with your class today with the Origami Organelles Frog Dissection Model.

With this model, your students can carry out a dissection using our detailed paper model of a frog. Use to prepare students for a dissection or as an alternative to a real dissection.

  • Students assemble the easy-to-make frog using the colorful parts working alone or in small groups.
  • Build the frog and then add the organs to the model.
  • The model can be used to identify the organs of a frog including the heart, lungs, liver, gall bladder, spleen, esophagus, stomach, pancreas, small intestine, large intestine, urinary bladder, kidneys, adrenal body, fat bodies, cloaca, male and female parts.
  • Can be used to look at different organ systems and for comparative anatomy.
  • Takes about 30 minutes for a group of students to make the model.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions:
38 cm x 27 cm (15 in. x 10 in.)

Download includes:
Instructions, color and mono model templates, and classroom PowerPoint presentation

All you need:
Paper, scissors, and tape

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology - Frog
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.