Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDVOTEK™ Origami Organelles Simple Plant Cell Model



Click to view available options
Grade:
4 to 12
Description
- Your students will enjoy learning about cells with this colorful model. The model covers all the main parts of plant cells (cell membrane, cell wall, chloroplasts, nucleus, chromosomes, mitochondria). For more advanced students, you can extend the model with optional parts (rough ER, smooth ER, and Golgi).
- Students make their cell model using the colorful parts working alone or in small groups. They can make a simple version or use the optional parts to show more features. It can be printed in color, or in mono on colored or white paper.
Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.
Dimensions:
18 cm x 14 cm (7 in. x 5 in.)
Download includes:
Instructions, model templates, and full color PowerPoint class presentation
All you need:
Paper, scissors, and tape
Specifications
Specifications
Grade | 4 to 12 |
Format | Model |
Languages | English |
Subject | Biology - Cells |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction