Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Soil Model
SDPSCI_ED

Catalog No. s05715 Shop All EDVOTEK Products
Click to view available options
Class Size:
75 Students
This item is not returnable. View return policy

Catalog No. S05715

Supplier: EDVOTEK™ EVT346

This item is not returnable. View return policy

Show the layers in soil with the 3 dimensional Origami Organelles Soil Model.

Soil covers nearly all of our planet's surface and is essential for plants to grow. Different types of soil are made by different types of parent rock. Show that soil forms layers and how these differ with our easy to make model.

For use in class or for distance learning at home.

  • First, students make their model of a soil profile using the colorful parts of the template.
  • Utilize the model it to look at the topsoil, subsoil and parent material.
  • Takes just 20 minutes to build the model.

The model can also be used to show more detailed layers (horizons) in a soil profile covering the O, A, E, B, C horizons.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Specifications

Grade 4 to 12
Includes 3D Model
Class Size 75 Students
Product Type Origami Organelles Soil Model
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.