Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chums™ Original Cotton Single Breakaway Eyewear Retainers

Catalog No. p-4099246 Shop All Chums Ltd Products
Encompass
Click to view available options
Product Type:
Breakaway Retainer
Material:
Cotton
Color:
Black
Navy
Red
Royal

Catalog No. 19-808-7036

Supplier: Chums™ 13002105

Only null left
Add to Cart
Add to Cart

Features a reconnecting single breakaway for situations where choking or snagging hazards exist

  • Fits most standard frames
  • Reconnecting breakaway pulls apart with six pounds of pressure
  • Field tested eyewear retainer
  • Available in a variety of colors
  • Logo option available
  • Made in the USA

Specifications

Color Navy
Product Type Breakaway Retainer
For Use With (Equipment) Standard frames
Includes Reconnecting single breakaway
Material Cotton
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.