Learn More
Description
World's first 100% cotton, fully adjustable eyewear retainer
Tough and durable, yet soft enough for everyday use.
- Fits most standard frames
- Logo option available
- Made in the USA
Order Info
Available in a variety of popular colors.
Specifications
Specifications
| Color | Black |
| Product Type | Eyewear Retainer |
| For Use With (Equipment) | Fits most standard frames |
| Material | Cotton |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.