Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Drummond™ Original Pipet-Aid™ Pipet Controller
Click to view available options
Model:
Complete (Dual Pump, TC Nosepiece), CE/Eur
Standard, CE/Eur
TC Nosepiece
Voltage:
110 V
220 V
Description
A patented, easy-to-use volume control mechanism permits precise fluid intake and delivery. The check valve protects against accidental fluid intake into the handle.
- Can be used with glass or plastic pipets
- Autoclavable components
- Optional EZ Grip Tissue Culture (TC) Nosepiece prevents cross-contamination due to over-pipetting
- Special dual pump filtration unit for use with biohazardous materials is available
- 220V CE models for use in Europe only (not for use in the U.S.A)
Order Info
Complete models include Pipet-Aid, TC nosepiece, four extra filters and dual-pump filtration unit. Contact your Fisher Scientific Customer Service Representative for available replacement parts.
Specifications
Specifications
Compatible Tips | Standard |
Autoclavable | Autoclavable |
Model | Standard, CE/Eur |
Type | Filler/Dispenser |
Volume (Metric) | 1 to 150 mL |
Filter Type | Standard |
For Use With (Application) | Pipetting of biohazardous materials |
For Use With (Equipment) | Glass or plastic pipets |
Voltage | 220 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction