Learn More
Thermo Scientific™ Orion™ 8115BNUWP ROSS Ultra™ Electrode PROMO
Description
Includes:
- 1 meter cable
Requires:
- pH buffers(pH 7 ±1 to 3 pH units) Beakers, plastic or glass
- Magnetic stirrer or Orion stirrer probe
- Distilled or deionized water
Compatible with:
BNWP connector is compatible with any standard BNC connector both on Thermo Scientific™ Orion Star™ Series meters and competitive meter models.
Recommended for:
- For use in clinical, pharmaceutical and food labs, wherever sample size is a constraint

Specifications
Specifications
Description | Epoxy body, semi-micro tip combination electrode; 165 x 6 mm |
Connector Types | BNC |
Junction | Glass Fiber |
Material | Epoxy Body |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 100°C |
Type | Electrode |
Filling Solution Type | 3M KCl |
Internal Reference | ROSS |
Precision | 0.01 |
Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.