Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Supplier: Thermo Scientific™ AC4070
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors
Specifications
Chlorine | |
Orion AQUAfast AQ4000 Colorimeter | |
30 Auto-ID Ampoules | |
Colorimetric water and wastewater testing equipment | |
0.4 to 5 mg/L Cl2 | |
30 Ampoules |
Ampoule | |
AQUAfast chlorine (free & total) Auto-ID ampoules, 30 pack | |
DPD | |
AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER) | |
Drinking Water: Standard Methods 4500-Cl G / ATP EPA-approved Orion Method AC4070; Wastewater: Standard Methods 4500-Cl G-2000 / ATP EPA-approved Orion Method AC4070 (total) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.