Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
$90.20 - $114.20
Catalog Number | Mfr. No. | Range | Type | Method | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Range | Type | Method | Price | Quantity & Availability | |||
13-644-932
|
Thermo Scientific™
AC4010 |
0.2 to 3 mg/L N | Ammonia | Salicylate |
Pack of 30 for $90.20
|
|
|||
13-650-117
|
Thermo Scientific™
AC4011 |
1.5 to 14 mg/L N | Ammonia | Direct Nesslerization |
Pack of 30 for $109.60
|
|
|||
13-650-116
|
Thermo Scientific™
AC4012 |
0.5 to 7 mg/L N | Ammonia | Direct Nesslerization |
Pack of 30 for $104.10
|
|
|||
13-642-175
|
Thermo Scientific™
AC4048 |
0.2 to 5 mg/L O3 | Ozone | DPD |
Each for $105.70
|
|
|||
13-642-161
|
Thermo Scientific™
AC4070 |
0.4 to 5 mg/L Cl2 | Chlorine | DPD |
Each for $114.20
|
|
|||
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.