Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis Spectrophotometers

Catalog No. p-7160112
Click to view available options
Type:
7100 Vis Spectrophotometer
8100 UV-Vis Spectrophotometer
Plug Type:
NA, EU, UK Power cord
Spectral Bandwidth:
2 nm
5 nm

Catalog No. 15-200-945

Supplier: Thermo Scientific™ AQ7100

Only null left
Add to Cart
Add to Cart
Request a Quote

Includes

Test Tube Holder for vials and test tube from 12 to 25 mm OD, AquaMate Water Analysis Methods Software, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362)

Optimized for your water analysis needs AquaMate 7100 Vis and AquaMate 8100 UV-Vis Spectrophotometers provide fast and reliable results

Thermo Scientific™ Orion™ AquaMate UV-Vis spectrophotometers include over 260 preprogrammed methods, and are designed to meet the needs of water and wastewater analysis laboratories. AquaMate UV-Vis and Visible-only spectrophotometers incorporate advanced features and functions for ultimate flexibility in operation.

Pre-programmed Methods:

  • More than 260 preprogrammed methods offer easy and convenient measurements using Orion AQUAfast™, Merck™ or CHEMetrics™ chemistries
  • Preprogrammed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
  • Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries

Versatility:

  • Glove friendly 7-inch touch screen controller
  • Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50mm rectangular vials and an adjustable round vial holder that accommodates vials and test tube from 12 to 25 mm OD

Verification:

  • Performance verification tests ensure wavelength accuracy and instrument functionality
  • Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP
  • Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards
  • Built-in test methods for stray light, noise and resolution are available to validate instrument performance further

Compliance

CE / TUV

Warranty and Services

1 Year

TRUSTED_SUSTAINABILITY

Specifications

Baseline Flatness ±0.005 A
Certifications/Compliance CE / TUV
Compartments Accessible from top, front or side Able to accommodate cells up to 100 mm cells Available accessory for test tubes up to 25 mm diameter and 150 mm height Removable, washable sample compartment liner with magnetic placement and hold-down
Connections Single USB-A
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Depth (Metric) 35.5 cm
Detector Type Dual Silicon Photodiodes
Display 7 in. High Definition Color Touchscreen, Fixed, 800 x 1280 pixels
Dimensions (L x W x H) 35.5 x 38.5 x 19.5 cm
Light Source Tungsten Halogen
Spectral Bandwidth 5 nm
Type 7100 Vis Spectrophotometer
Wavelength Range 325 to 1100 nm
Drift <0.0010 A/hr. (At 500 nm after 1 hour warmup)
Electrical Requirements External AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz
For Use With (Application) Water and Waste Water Analysis
Height (Metric) 19.5 cm
Includes
Test Tube Holder for vials and test tube from 12 to 25 mm OD, AquaMate Water Analysis Methods Software, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362).
Interface Glove friendly touch-screen
Keypad Glove- friendly touchscreen
Lamp Life >1000 hrs.
Length (Metric) 35.5 cm
Noise (RMS at 500 nm 60 consecutive measurements) ≤0.00020A at 0A at 260 and 500 nm ≤0.00030A at 1A at 260 and 500 nm ≤0.00040A at 2A at 260 and 500 nm
Optical Design Double Beam
Photometric Accuracy ±0.002 A at 0.5 A, ±0.004 A at 1.0 A, ±0.008 A at 2.0 A
Photometric Display 3A to +5A
Photometric Range –3A to +3.5A
Photometric Repeatability ±0.001A at 1A measured at 1.0A at 546 nm
Plug Type NA, EU, UK Power cord
Printer Included Snap-on printer available
Scan Speed Automatic – up to 1800 nm/min.
Stray Light <0.05%T at 340 nm and 400 nm
Warranty 1 Year
Wavelength Accuracy ±0.5 nm
Wavelength Data Interval 0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability < ±0.2 nm
Weight (Metric) 7.5 kg
Width (Metric) 38.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.