Learn More
Thermo Scientific™ Orion™ Calcium Ion Selective Electrodes
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
None
For Use With (Equipment):
Combination Calcium Electrode
Half-Cell Calcium Electrode
ISE Meter, Module 972001
Orion 900100, Module 932001
Type:
Combination Calcium Electrode
Half-Cell Calcium Electrode
Replacement Module
Description
Measure calcium in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Calcium Electrodes.
A separate reference electrode is unnecessary with this electrode, making it convenient to use with small samples.
Related Products:
- Calibration standards: 922006 (0.1 M CaCl2); 923206 (100ppm)
- Required ISA: 932011
Specifications
Specifications
Connector Types | BNC |
Description | Half-Cell Calcium Electrode with BNC Connector |
Junction | Sure-Flow Junction |
For Use With (Equipment) | Orion 900100, Module 932001 |
Type | Half-Cell Calcium Electrode |