Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Orion™ Conductivity Conditioning Solution; 5 x 60 mL bottles
Click to view available options
:
Conductivity conditioning solution; 5 x 60mL bottles
Description
Perform virtually any electrochemistry analysis with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Choose from a complete selection of conductivity accessories including conductivity standards and TDS (total dissolved solids) standards. All conductivity standards are traceable to NIST™ standard reference materials.
Conductivity Standards
- Traceable to NIST standard reference materials
- Designed to meet all end user requirements, including laboratory grade and DIN standards
- Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature

Specifications
Specifications
Description | Conductivity Conditioning Solution |
Packaging | Bottle |
Traceability to NIST | Yes |
Specific Gravity | 980 kg/m3 |
Conductivity | — |
Color | Light Green |
pH | 11.5 |
Boiling Point | 100°C |
For Use With (Equipment) | Conductivity Probes (011050MD, 011050A and 011050) |
Solubility | Soluble |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction