Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Ion Specific Electrode Kits

Catalog No. 13301145
Click to view available options
Electrode:
Ammonia ISE
Fluoride ISE
HP Ammonia ISE

Catalog No. 13-301-145

Supplier: Thermo Scientific™ 9609BNSLN

Ensure correct measurement by using high quality electrodes and reagents for ion specific measurement. Ordering is easy! All the consumables you need to measure fluoride or ammonia in water and wastewater are available with one part number. Now with NEW 0.5 ppm and 5.0 ppm Fluoride Standards with TISAB II for low level fluoride analysis

Perform quick and accurate measurements by using high quality Thermo Scientific™ Orion™ Ion Specific Electrodes and reagents for ion specific measurement. Ordering is easy! We offer a comprehensive portfolio of ion selective electrode calibration standards, ionic strength adjusters, reagents, and fill solutions for your ISE measurement needs. All the consumables you need to measure fluoride or ammonia in water and wastewater are available with one part number. Now with NEW 0.5 ppm and 5.0 ppm Fluoride Standards with TISAB II for low level fluoride analysis.

Fluoride ISE Kit

  • Orion Combination Fluoride ISE features the Thermo™ Scientific™ Orion™ Sure-Flow™ junction which contributes to highly stable and precise fluoride measurements while simplifying electrode maintenance.
  • 0.5, 1.0, 2.0, 5.0 and 10.0 ppm Fluoride Standards are pre-mixed with total ionic strength adjustor for your convenience. No dilution and no addition of ionic strength adjustor needed.
  • Total Ionic Strength Adjustor (TISAB II) included for addition to samples

Ammonia ISE Kit

  • This kit includes the Orion Standard Performance Ammonia ISE (Cat. No. 9512BNWP) and is ideal for mid to high ammonia level measurements.
  • The Ionic Strength Adjuster (ISA) (Cat. No. 951211) included in the kit, is formulated for samples containing metallic ions and has pH indicating blue dye added, so you know the pH is correct for analysis.
  • This kit includes the Orion Standard Performance Ammonia ISE (Cat. No. 9512BNWP) and is ideal for mid to high ammonia level measurements.
  • The Ionic Strength Adjuster (ISA) (Cat. No. 951211) included in the kit, is formulated for samples containing metallic ions and has pH indicating blue dye added, so you know the pH is correct for analysis.
TRUSTED_SUSTAINABILITY

Specifications

Applications Fluoride Analysis
Connector Types BNC (Waterproof)
Description Fluoride Ion Selective Electrode and Reagent Kit
Diameter (English) Body 0.55 in.
Diameter (Metric) Body 14mm
Diameter (Metric) Cap 17mm
Filling Solution Type Optimum Results A fill sol PN 90061
Internal Reference 9606BNWP
Junction Sure-Flow Junction
Length (English) 4.75 in.
Length (English) Cable 39.40 in.
Length (Metric) 110mm
Length (Metric) Cable 1m
Material Epoxy
Operating Range (Metric) RANGES: 1 x 10E-6M to saturated
Temperature (English) Operating 32-180°F
Membrane pellet
Model 9606BNWP
Electrode Fluoride ISE
Temperature (Metric) Range 0-80°C
For Use With (Equipment) ISE Meter
Temperature (Metric) Operating 25°C
Type Fluoride Ion Selective Electrode and Reagent Kit
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.