Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions

Catalog No. p-4521260
Click to view available options
Quantity:
60 mL
475 mL
Chemical Name or Material:
Hyamine 1622 Titrant
Sodium Lauryl Sulfate Standard
Surfactant Reagent, Nonionic

Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).

Use Thermo Scientific™ Orion™ ISE Surfactant Analysis Solutions for titrations with surfactant ion selective electrodes (ISE).

All surfactant analysis solutions are packaged as one 1 pint (475mL) bottle, except Cat. No. 654202 packaged as one 2 oz. (60mL) bottle.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Lauryl Sulfate Standard
For Use With (Application) PH ion conductivity & oxygen measurement
For Use With (Equipment) Surfactant Ion Selective Electrode
Quantity 60 mL
Solution Type Surfactant Analysis
Packaging Bottle

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.