Learn More
Thermo Scientific™ Orion Lab Star Bench Meter Accessories
Catalog No. p-7224958
Click to view available options
Type:
Dust Cover
Electrode Stand
Power Adapter
USB Cable
This item is not returnable. View return policy
This item is not returnable. View return policy
Orion Lab Star series meter accessories support your Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter or Orion Lab Star DO113 DO meter.
Select Orion Lab Star series meter accessories to support your Orion Lab Star PH111 pH/mV meter, Orion Lab Star EC112 conductivity meter or Orion Lab Star DO113 dissolved oxygen meter. Utilize the meter dust cover accessory for extended meter storage. Select a replacement meter-attached electrode holder, universal power adapter or computer cable when needed. The Orion Lab Star series pH meter, conductivity meter and DO meter are ideal for simplified, budget-friendly lab testing and data recording.
Orion Lab Star Series Meter Accessories Features
  • LSTAR-ARM Orion Lab Star series meter-attached electrode stand allows you to replace your electrode stand as needed and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
  • LSTAR-PWR Orion Lab Star series universal power supply, 100-240V 50/60 Hz, allows you to replace your power adapter as needed and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
  • LSTAR-USB Orion Lab Star series USB computer cable allows you to replace your computer cable as needed and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
  • LSTAR-CVR Orion Lab Star series meter dust cover protects your meter from dust and debris while in storage and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
Additional Accessories Compatible with Orion Lab Star Series Meters
  • STARA-106 ink-ribbon compact printer, 100-240V 50/60 Hz
TRUSTED_SUSTAINABILITY
Specifications
Description Orion Lab Star series electrode stand with meter-attachable bracket
Type Electrode Stand
Certifications/Compliance None
Compatibility Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.