Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Orion Lab Star PH111 Bench pH Meters PROMO
Click to view available options
Model:
Difficult Sample Kit Option
Standard Kit Option
Description
Orion Lab Star PH111 pH Meter Features
- Budget-friendly pH meter for simplified pH and ORP testing
- Display shows pH, mV and temperature simultaneously for accurate data recording
- Measure -2.000 to 18.000 pH, -2000.0 to +2000.0 mV and -5.0 to 105.0°C or 23.0 to 221.0°F to ensure comprehensive measurement capabilities
- Perform one to five point pH calibrations, one point ORP calibrations and one point temperature adjustment calibrations for proficient SOP compliance
- Large 5 inch backlit display makes viewing and recording measurements easy
- Small meter footprint saves valuable bench space in your lab
- Onscreen stabilizing and ready indicators help identify when readings are complete
- pH electrode condition icon helps ensure reliable electrode performance
- Date and time data stamped data and calibration logs support GLP compliance
- View active pH, ORP and temperature calibration logs with date and time stamp to ensure accurate, reproducible measurements
- Use 500-point data log with date and time stamp to record key measurements
- Output data and calibration logs to a printer or computer for long-term data storage
Meter and Meter Kits
- LSTAR1110 is the pH meter and stand option, which allows you to purchase your pH electrode separately
- LSTAR1115 is the standard pH meter kit option, for common pH samples such as drinking water, tap water, buffers (non-TRIS) and general purpose student use
- LSTAR1116 is the easy-to-clean bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included refillable pH electrode is easy-to-clean and restore
- LSTAR1117 is the low-maintenance bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included gel-filled pH electrode is low-maintenance and easy to operate
- LSTAR1118 is the difficult sample pH meter kit option, for pH samples that are thick, viscous or semi-solid including foods, beverages, wastewaters, emulsions, soils, slurries, suspended solids or sludges
Specifications
Specifications
| Description | Orion Lab Star PH111 bench pH/mV meter standard kit |
| Includes | 9157 BNMD refillable epoxy-body pH/ATC electrode, 916099 pH 4/7/10 buffer, meter with electrode stand, PC cable, universal power adapter |
| Data Management Details | Transfer single or all data points to printer or computer |
| Outputs | Computer or printer (STARA-106) |
| Accuracy (pH) | ±0.005, ± 1 LSD |
| Accuracy (Temperature) | ±0.3°C/±0.5°F |
| Accuracy (mV) | ±0.2 or ±0.05%, whichever is greater |
| For Use With (Application) | Standard pH Meter Kit Option, for Common pH Samples Such as Drinking Water, Tap Water, Buffers (Non-TRIS) and General Purpose Student Use |
| Measurement Modes | pH with mV and Temperature, ORP with mV and Temperature |
| Calibration | pH, ORP, Temperature |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction